2,4,5-Trifluorobenzyl bromide 97%

Supplier: Apollo Scientific
Danger

PC7285S-100G PC7285S-25G PC7285S-500G PC7285S-1KG
APOSPC7285S-100GEA 203 EUR
APOSPC7285S-100G APOSPC7285S-25G APOSPC7285S-500G APOSPC7285S-1KG
2,4,5-Trifluorobenzyl bromide 97%
2,4,5-Trifluorobenzyl bromide
Formula: C₇H₄BrF₃
Density: 1,71 g/cm³ (25 °C)
Flash Pt: 90,6 °C
Storage Temperature: Ambient
MDL Number: MFCD00061209
CAS Number: 157911-56-3
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR