4-Bromo-1-fluoro-2-(trifluoromethoxy)benzene 99%

Supplier: Apollo Scientific
Warning

PC7291-1G PC7291-5G PC7291-250MG PC7291-25G
APOSPC7291-1GEA 56.7 EUR
APOSPC7291-1G APOSPC7291-5G APOSPC7291-250MG APOSPC7291-25G
4-Bromo-1-fluoro-2-(trifluoromethoxy)benzene 99%
4-Bromo-1-fluoro-2-(trifluoromethoxy)benzene
Formula: C₇H₃BrF₄O
Storage Temperature: Ambient
MDL Number: MFCD06660160
CAS Number: 886496-45-3
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR