2,2,2-Trifluoroethyl toluene-4-sulphonate 98%

Supplier: Apollo Scientific
Warning

PC7376-100G PC7376-500G PC7376-1KG
APOSPC7376-100GEA 37.7 EUR
APOSPC7376-100G APOSPC7376-500G APOSPC7376-1KG
2,2,2-Trifluoroethyl toluene-4-sulphonate 98%
2,2,2-Trifluoroethyl toluene-4-sulphonate

Useful reagent for introduction of trifluoroethyl group.

Formula: C₉H₉F₃O₃S
Boiling Pt: 87…92 °C/0,1mmHg
Melting Pt: 36…38 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000443
CAS Number: 433-06-7
EINECS: 207-085-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR