4-Bromo-2-fluorobenzyl alcohol 98+%

Supplier: Apollo Scientific
Warning

PC7381-100G PC7381-25G PC7381-5G
APOSPC7381-100GEA 108 EUR
APOSPC7381-100G APOSPC7381-25G APOSPC7381-5G
4-Bromo-2-fluorobenzyl alcohol 98+%
4-Bromo-2-fluorobenzyl alcohol
Formula: C₇H₆BrFO
Melting Pt: 30…35 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00143265
CAS Number: 188582-62-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR