4-(Trifluoromethoxy)benzyl mercaptan 97%

Supplier: Apollo Scientific
Warning

Synonyms: 4-(Trifluoromethoxy)benzylthiol

PC7438UL-250MG
APOSPC7438UL-250MGEA 23.4 EUR
APOSPC7438UL-250MG
4-(Trifluoromethoxy)benzyl mercaptan 97%
4-(Trifluoromethoxy)benzyl mercaptan
Formula: C₈H₇F₃OS
Boiling Pt: 93…95 °C/15mmHg
Storage Temperature: Ambient
MDL Number: MFCD00052327
CAS Number: 175278-03-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR