4-(Trifluoromethoxy)phenylacetonitrile 98%

Supplier: Apollo Scientific
Warning

PC7440H-100G PC7440H-25G PC7440H-5G
APOSPC7440H-100GEA 128 EUR
APOSPC7440H-100G APOSPC7440H-25G APOSPC7440H-5G
4-(Trifluoromethoxy)phenylacetonitrile 98%
4-(Trifluoromethoxy)phenylacetonitrile
Formula: C₉H₆F₃NO
Boiling Pt: 96 °C/3,75mmHg
Melting Pt: 28…33 °C
Density: 1,28 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00061240
CAS Number: 49561-96-8
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR