3-(Trifluoromethyl)benzamidoxime 97%

Supplier: Apollo Scientific
Warning

Synonyms: N'-Hydroxy-3-(trifluoromethyl)-benzenecarboximidamide, N-Hydroxy-3-(trifluoromethyl)benzimidamide

PC7507L-100G PC7507L-25G PC7507L-5G
APOSPC7507L-100GEA 192 EUR
APOSPC7507L-100G APOSPC7507L-25G APOSPC7507L-5G
3-(Trifluoromethyl)benzamidoxime 97%
3-(Trifluoromethyl)benzamidoxime
Formula: C₈H₇F₃N₂O
MW: 204.15 g/mol
Melting Pt: 85…88 °C
Storage Temperature: Ambient
MDL Number: MFCD02647312
CAS Number: 40067-80-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR