4-(Trifluoromethyl)benzamidoxime 95%

Supplier: Apollo Scientific
Warning

PC7507M-1G PC7507M-5G
APOSPC7507M-1GEA 21.9 EUR
APOSPC7507M-1G APOSPC7507M-5G
4-(Trifluoromethyl)benzamidoxime 95%
4-(Trifluoromethyl)benzamidoxime
Formula: C₈H₇F₃N₂O
MW: 204.15 g/mol
Melting Pt: 121…128 °C
Density: 0.936 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00053042
CAS Number: 22179-86-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR