4-(Methylsulphonyl)-2-(trifluoromethyl)benzyl bromide 98%

Supplier: Apollo Scientific
Danger

PC7521-1G PC7521-5G PC7521-10G
APOSPC7521-1GEA 290 EUR
APOSPC7521-1G APOSPC7521-5G APOSPC7521-10G
4-(Methylsulphonyl)-2-(trifluoromethyl)benzyl bromide 98%
4-(Methylsulphonyl)-2-(trifluoromethyl)benzyl bromide
Formula: C₉H₈BrF₃O₂S
Melting Pt: 121…123 °C
Storage Temperature: Ambient
MDL Number: MFCD16606069
CAS Number: 934557-65-0
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR