2-(Trifluoromethyl)benzophenone 98%

Supplier: Apollo Scientific
Warning

PC7568-100G PC7568-25G PC7568-5G
APOSPC7568-100GEA 410 EUR
APOSPC7568-100G APOSPC7568-25G APOSPC7568-5G
2-(Trifluoromethyl)benzophenone 98%
2-(Trifluoromethyl)benzophenone
Formula: C₁₄H₉F₃O
Melting Pt: 58…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00000377
CAS Number: 727-99-1
EINECS: 211-972-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR