4-(Trifluoromethyl)benzophenone 97%

Supplier: Apollo Scientific
Warning

PC7570-100G PC7570-25G PC7570-5G
APOSPC7570-100GEA 275 EUR
APOSPC7570-100G APOSPC7570-25G APOSPC7570-5G
4-(Trifluoromethyl)benzophenone 97%
4-(Trifluoromethyl)benzophenone
Formula: C₁₄H₉F₃O
Melting Pt: 114…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00000400
CAS Number: 728-86-9
EINECS: 211-974-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR