4-Bromo-2-fluoro-6-methoxybenzeneboronic acid 95+%

Supplier: Apollo Scientific
Warning

PC7585-1G PC7585-5G
APOSPC7585-1GEA 68.3 EUR
APOSPC7585-1G APOSPC7585-5G
4-Bromo-2-fluoro-6-methoxybenzeneboronic acid 95+%
4-Bromo-2-fluoro-6-methoxybenzeneboronic acid
Formula: C₇H₇BBrFO₃
Storage Temperature: Refrigerator
MDL Number: MFCD09475882
CAS Number: 957035-32-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR