3-Bromo-4-fluorobenzamide

Supplier: Apollo Scientific
Warning

PC7598-250MG
APOSPC7598-250MGEA 21.9 EUR
APOSPC7598-250MG
3-Bromo-4-fluorobenzamide
3-Bromo-4-fluorobenzamide
Formula: C₇H₅BrFNO
Melting Pt: 126…127 °C
Storage Temperature: Ambient
MDL Number: MFCD03094431
CAS Number: 455-85-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR