Ethyl-3-chloro-5-(trifluoromethyl)picolinate

Supplier: Apollo Scientific
Warning

Synonyms: Ethyl 3-chloro-5-(trifluoromethyl)picolinate, Ethyl 3-chloro-5-(trifluoromethyl)pyridine-2-carboxylate

PC7620-500MG
APOSPC7620-500MGEA 21.9 EUR
APOSPC7620-500MG
Ethyl-3-chloro-5-(trifluoromethyl)picolinate
Ethyl 3-chloro-5-(trifluoromethyl)picolinate
Formula: C₉H₇ClF₃NO₂
MW: 253.60 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06656414
CAS Number: 128073-16-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR