trans-2-(Trifluoromethyl)cinnamic acid 97%

Supplier: Apollo Scientific
Warning

Synonyms: (2E)-3-[2-(Trifluoromethyl)phenyl]prop-2-enoic acid, (E)-3-[2-(Trifluoromethyl)phenyl]acrylic acid, (E)-3-(2-(Trifluoromethyl)phenyl)acrylic acid

PC7662-10G
APOSPC7662-10GEA 39.3 EUR
APOSPC7662-10G
trans-2-(Trifluoromethyl)cinnamic acid 97%
trans-2-(Trifluoromethyl)cinnamic acid
Formula: C₁₀H₇F₃O₂
Melting Pt: 199 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004381
CAS Number: 98386-81-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR