2-(Trifluoromethyl)thiophenol 97%

Supplier: Apollo Scientific
Warning

Synonyms: 2-(Trifluoromethyl)benzene-1-thiol, 2-(Trifluoromethyl)benzenethiol, 2-Mercaptobenzotrifluoride

PC7771-100G PC7771-25G PC7771-5G
APOSPC7771-100GEA 522 EUR
APOSPC7771-100G APOSPC7771-25G APOSPC7771-5G
2-(Trifluoromethyl)thiophenol 97%
2-(Trifluoromethyl)thiophenol
Formula: C₇H₅F₃S
Boiling Pt: 175…177 °C
Density: 1,35 g/cm³ (25 °C)
Flash Pt: 68 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00068896
CAS Number: 13333-97-6
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR