(2,4,6-Trifluorophenyl)acetonitrile 97%

Supplier: Apollo Scientific
Warning

PC7818N-1G PC7818N-25G PC7818N-5G
APOSPC7818N-1GEA 21.9 EUR
APOSPC7818N-1G APOSPC7818N-25G APOSPC7818N-5G
(2,4,6-Trifluorophenyl)acetonitrile 97%
(2,4,6-Trifluorophenyl)acetonitrile
Formula: C₈H₄F₃N
Boiling Pt: 103 °C/3mmHg
Storage Temperature: Ambient
MDL Number: MFCD00061221
CAS Number: 220227-80-5
UN: 3276
ADR: 6.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR