Zinc fluoride, anhydrous 99%

Supplier: Apollo Scientific
Danger

PC8030-100G PC8030-250G PC8030-25G
APOSPC8030-100GEA 90.5 EUR
APOSPC8030-100G APOSPC8030-250G APOSPC8030-25G
Zinc fluoride, anhydrous 99%
Zinc fluoride
Formula: F₂Zn
Boiling Pt: 1500 °C
Melting Pt: 872 °C
Density: 4,95 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011298
CAS Number: 7783-49-5
EINECS: 232-001-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR