2-Bromo-1,3-dimethyl-5-fluorobenzene 98%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Bromo-5-fluoro-m-xylene, 2-Bromo-5-fluoro-1,3-dimethylbenzene

PC8087-10G PC8087-25G PC8087-5G
APOSPC8087-10GEA 59.6 EUR
APOSPC8087-10G APOSPC8087-25G APOSPC8087-5G
2-Bromo-1,3-dimethyl-5-fluorobenzene 98%
2-Bromo-1,3-dimethyl-5-fluorobenzene
Formula: C₈H₈BrF
MW: 203.052 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09864702
CAS Number: 14659-58-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR