3-Fluoro-p-anisidine

Supplier: Apollo Scientific
Warning

PC8204-25G PC8204-5G
APOSPC8204-25GEA 33.5 EUR
APOSPC8204-25G APOSPC8204-5G
3-Fluoro-p-anisidine
3-Fluoro-p-anisidine
Formula: C₇H₈FNO
Boiling Pt: 135 °C/18mmHg
Melting Pt: 80…84 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00075040
CAS Number: 366-99-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR