4-(Trifluoromethyl)anisole

Supplier: Apollo Scientific
Warning

PC8229-5G PC8229-1G PC8229-25G
APOSPC8229-5GEA 31.9 EUR
APOSPC8229-5G APOSPC8229-1G APOSPC8229-25G
4-(Trifluoromethyl)anisole
4-(Trifluoromethyl)anisole
Formula: C₈H₇F₃O
MW: 176.14 g/mol
Boiling Pt: 166…170 °C
Melting Pt: –9 °C
Density: 1.2445 g/cm³ (25 °C)
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD03094456
CAS Number: 402-52-8
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR