α-Bromo-4-chloro-2,6-difluorotoluene 98%

Supplier: Apollo Scientific
Danger

PC8242-10G PC8242-5G PC8242-1G PC8242-25G
APOSPC8242-10GEA 282 EUR
APOSPC8242-10G APOSPC8242-5G APOSPC8242-1G APOSPC8242-25G
α-Bromo-4-chloro-2,6-difluorotoluene 98%
α-Bromo-4-chloro-2,6-difluorotoluene
Formula: C₇H₄BrClF₂
Storage Temperature: Ambient
MDL Number: MFCD03094463
CAS Number: 537013-52-8
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR