3,5-Difluoro-4-pyridinamine 97+%

Supplier: Apollo Scientific
Warning

PC8304-100MG PC8304-1G PC8304-500MG
APOSPC8304-100MGEA 21.9 EUR
APOSPC8304-100MG APOSPC8304-1G APOSPC8304-500MG
3,5-Difluoro-4-pyridinamine 97+%
3,5-Difluoro-4-pyridinamine
Formula: C₅H₄F₂N₂
Melting Pt: 99…101 °C
Storage Temperature: Ambient
MDL Number: MFCD01862101
CAS Number: 159783-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR