N-Benzyl-2-bromo-5-(trifluoromethoxy)benzenesulphonamide

Supplier: Apollo Scientific

PC8319-1G
APOSPC8319-1GEA 212 EUR
APOSPC8319-1G
N-Benzyl-2-bromo-5-(trifluoromethoxy)benzenesulphonamide
N-Benzyl 2-bromo-5-(trifluoromethoxy)benzenesulphonamide
Formula: C₁₄H₁₁BrF₃NO₃S
Storage Temperature: Ambient
MDL Number: MFCD09800993
CAS Number: 951885-49-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR