Bromotrispyrrolidinophosphonium hexafluorophosphate

Supplier: Apollo Scientific
Danger

PC8353-100G PC8353-250G PC8353-25G
APOSPC8353-100GEA 200 EUR
APOSPC8353-100G APOSPC8353-250G APOSPC8353-25G
Bromotrispyrrolidinophosphonium hexafluorophosphate
Bromo-tris-pyrrolidinophosphonium hexafluorophosphate
Formula: C₁₂H₂₄BrF₆N₃P₂
Melting Pt: 139 °C
Storage Temperature: Freezer
MDL Number: MFCD00077412
CAS Number: 132705-51-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR