6-fluoro-4-methylpyridin-3-amine 97%

Supplier: Apollo Scientific
Danger

Synonyms: 6-Fluoro-4-methylpyridin-3-amine, 5-Amino-2-fluoro-4-methylpyridine

PC8384-1G PC8384-25G
APOSPC8384-1GEA 34.8 EUR
APOSPC8384-1G APOSPC8384-25G
6-fluoro-4-methylpyridin-3-amine 97%
6-fluoro-4-methylpyridin-3-amine
Formula: C₆H₇FN₂
MW: 126.134 g/mol
Melting Pt: 57…62 °C
Storage Temperature: Ambient
MDL Number: MFCD09864696
CAS Number: 954236-33-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR