2-[(4-Fluorobenzyl)sulphanyl]ethylamine hydrochloride

Supplier: Apollo Scientific
Warning

PC8586-1G PC8586-250MG
APOSPC8586-1GEA 205 EUR
APOSPC8586-1G APOSPC8586-250MG
2-[(4-Fluorobenzyl)sulphanyl]ethylamine hydrochloride
2-[(4-Fluorobenzyl)sulphanyl]ethylamine hydrochloride
Formula: C₉H₁₃ClFNS
Melting Pt: 167…169 °C
Storage Temperature: Ambient
MDL Number: MFCD09027133
CAS Number: 1170404-43-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR