Methyl-4-amino-3-fluorobenzoate

Supplier: Apollo Scientific
Warning

PC8633-25G PC8633-5G
APOSPC8633-25GEA 49.3 EUR
APOSPC8633-25G APOSPC8633-5G
Methyl-4-amino-3-fluorobenzoate
Methyl 4-amino-3-fluorobenzoate
Formula: C₈H₈FNO₂
Melting Pt: 107…110 °C
Storage Temperature: Ambient
MDL Number: MFCD08689703
CAS Number: 185629-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR