1-n-Butoxy-2,3-difluorobenzene 99%

Supplier: Apollo Scientific
Warning

PC8738-100G PC8738-250G PC8738-25G PC8738-500G
APOSPC8738-100GEA 87 EUR
APOSPC8738-100G APOSPC8738-250G APOSPC8738-25G APOSPC8738-500G
1-n-Butoxy-2,3-difluorobenzene 99%
1-n-Butoxy-2,3-difluorobenzene
Formula: C₁₀H₁₂F₂O
Storage Temperature: Ambient
MDL Number: MFCD07368738
CAS Number: 136239-66-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR