2-methyl-4-(trifluoromethoxy)phenol 97%

Supplier: Apollo Scientific
Warning

PC8770-1G PC8770-250MG PC8770-5G
APOSPC8770-1GEA 24.8 EUR
APOSPC8770-1G APOSPC8770-250MG APOSPC8770-5G
2-methyl-4-(trifluoromethoxy)phenol 97%
2-methyl-4-(trifluoromethoxy)phenol
Formula: C₈H₇F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD04972927
CAS Number: 129676-67-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR