4-(Aminomethyl)-3-fluoropyridine dihydrochloride

Supplier: Apollo Scientific
Danger

Synonyms: 3-Fluoroisonicotinylamine dihydrochloride, (3-Fluoropyridin-4-yl)methylamine dihydrochloride, (3-Fluoropyridin-4-yl)methanamine dihydrochloride

PC8935-250MG PC8935-500MG
APOSPC8935-250MGEA 418 EUR
APOSPC8935-250MG APOSPC8935-500MG
4-(Aminomethyl)-3-fluoropyridine dihydrochloride
4-(Aminomethyl)-3-fluoropyridine dihydrochloride
Formula: C₆H₇FN₂·2ClH
MW: 199.053 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12922842
CAS Number: 1257535-26-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR