3-Chloro-5-fluoropyridine-N-oxide 96%

Supplier: Apollo Scientific

PC900557-1G
APOSPC900557-1GEA 270 EUR
APOSPC900557-1G
3-Chloro-5-fluoropyridine-N-oxide 96%
3-Chloro-5-fluoropyridine N-oxide
Formula: C₅H₃ClFNO
Storage Temperature: Ambient
MDL Number: MFCD16036157
CAS Number: 1221793-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR