4-Amino-2-ethoxy-5-fluorobenzonitrile 96%

Supplier: Apollo Scientific

PC901587-1G
APOSPC901587-1GEA 270 EUR
APOSPC901587-1G
4-Amino-2-ethoxy-5-fluorobenzonitrile 96%
4-Amino-2-ethoxy-5-fluorobenzonitrile
Formula: C₉H₉FN₂O
MW: 180,179 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22689093
CAS Number: 1420800-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR