4-Methyl-3-(trifluoromethyl)phenylboronic acid 95%

Supplier: Apollo Scientific
Warning

PC901644-1G PC901644-5G
APOSPC901644-1GEA 243 EUR
APOSPC901644-1G APOSPC901644-5G
4-Methyl-3-(trifluoromethyl)phenylboronic acid 95%
4-Methyl-3-(trifluoromethyl)phenylboronic acid
Formula: C₈H₈BF₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD11042431
CAS Number: 947533-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR