Ethyl 5-amino-2-fluorobenzoate 95%

Supplier: Apollo Scientific

PC903640-5G
APOSPC903640-5GEA 65.4 EUR
APOSPC903640-5G
Ethyl 5-amino-2-fluorobenzoate 95%
Ethyl 5-amino-2-fluorobenzoate
Formula: C₉H₁₀FNO₂
MW: 183.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09037896
CAS Number: 123207-39-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR