2-[(Ethylamino)methyl]-4-fluorophenol 95%

Supplier: Apollo Scientific

PC904176-1G PC904176-250MG
APOSPC904176-1GEA 435 EUR
APOSPC904176-1G APOSPC904176-250MG
2-[(Ethylamino)methyl]-4-fluorophenol 95%
2-[(Ethylamino)methyl]-4-fluorophenol
Formula: C₉H₁₂FNO
Storage Temperature: Ambient
MDL Number: MFCD21641326
CAS Number: 1363166-27-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR