3-(4-Fluorophenoxy)propan-1-ol 95%

Supplier: Apollo Scientific

PC905206-1G
APOSPC905206-1GEA 574 EUR
APOSPC905206-1G
3-(4-Fluorophenoxy)propan-1-ol 95%
3-(4-Fluorophenoxy)propan-1-ol
Formula: C₉H₁₁FO₂
Storage Temperature: Ambient
MDL Number: MFCD08442180
CAS Number: 104413-57-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR