4-(Trifluoromethyl)cyclohexanemethanol (cis- and trans- mixture) 95%

Supplier: Apollo Scientific

PC906474-1G PC906474-5G
APOSPC906474-1GEA 335 EUR
APOSPC906474-1G APOSPC906474-5G
4-(Trifluoromethyl)cyclohexanemethanol (cis- and trans- mixture) 95%
4-(Trifluoromethyl)cyclohexanemethanol (cis- and trans- mixture)
Formula: C₈H₁₃F₃O
CAS Number: 883731-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR