2-Chloro-4-fluoro-6-nitroaniline 96%

Supplier: Apollo Scientific

PC909152-5G
APOSPC909152-5GEA 116 EUR
APOSPC909152-5G
2-Chloro-4-fluoro-6-nitroaniline 96%
2-Chloro-4-fluoro-6-nitroaniline
Formula: C₆H₄ClFN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD22571067
CAS Number: 153505-32-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR