(2,4-Difluorophenoxy)acetic acid 98%

Supplier: Apollo Scientific

PC911112-1G PC911112-5G
APOSPC911112-1GEA 190 EUR
APOSPC911112-1G APOSPC911112-5G
(2,4-Difluorophenoxy)acetic acid 98%
(2,4-Difluorophenoxy)acetic acid
Formula: C₈H₆F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD03422203
CAS Number: 399-44-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR