7-Fluoro-1,2,3,4-tetrahydronaphthalen-2-amine 95%

Supplier: Apollo Scientific
Warning

PC911133-100MG PC911133-250MG
APOSPC911133-100MGEA 335 EUR
APOSPC911133-100MG APOSPC911133-250MG
7-Fluoro-1,2,3,4-tetrahydronaphthalen-2-amine 95%
7-Fluoro-1,2,3,4-tetrahydronaphthalen-2-amine
Formula: C₁₀H₁₂FN
Storage Temperature: Refrigerator
MDL Number: MFCD06739166
CAS Number: 173998-63-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR