N,N-Diethyl chlorofluoroacetamide 97%

Supplier: Apollo Scientific
Warning

PC9117-25G PC9117-5G
APOSPC9117-25GEA 868 EUR
APOSPC9117-25G APOSPC9117-5G
N,N-Diethyl chlorofluoroacetamide 97%
N,N-Diethyl chlorofluoroacetamide
Formula: C₆H₁₁ClFNO
Boiling Pt: 113…115 °C/27mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD00101818
CAS Number: 364-94-3
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR