2-Bromo-4-(trifluoromethyl)phenylboronic acid

Supplier: Apollo Scientific
Warning

PC9160-1G PC9160-5G
APOSPC9160-1GEA 58 EUR
APOSPC9160-1G APOSPC9160-5G
2-Bromo-4-(trifluoromethyl)phenylboronic acid
2-Bromo-4-(trifluoromethyl)phenylboronic acid
Formula: C₇H₅BBrF₃O₂
Storage Temperature: Freezer
MDL Number: MFCD08532500
CAS Number: 959997-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR