2-Fluoro-6-methylbenzyl bromide

Supplier: Apollo Scientific
Danger

PC9288-1G PC9288-5G
APOSPC9288-1GEA 192 EUR
APOSPC9288-1G APOSPC9288-5G
2-Fluoro-6-methylbenzyl bromide
2-Fluoro-6-methylbenzyl bromide
Formula: C₈H₈BrF
Storage Temperature: Ambient
MDL Number: MFCD06660281
CAS Number: 886502-18-7
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR