3-{[3-(Trifluoromethyl)benzyl]sulphanyl}-1H-1,2,4-triazol-5-amine 97%

Supplier: Apollo Scientific

PC9492-1G PC9492-500MG PC9492-5G
APOSPC9492-1GEA 275 EUR
APOSPC9492-1G APOSPC9492-500MG APOSPC9492-5G
3-{[3-(Trifluoromethyl)benzyl]sulphanyl}-1H-1,2,4-triazol-5-amine 97%
3-{[3-(Trifluoromethyl)benzyl]sulphanyl}-1H-1,2,4-triazol-5-amine
Formula: C₁₀H₉F₃N₄S
Melting Pt: 136…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00141527
CAS Number: 303150-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR