(ɑ,ɑ,ɑ,2,3,5,6-Heptafluoro-p-tolyl)hydrazine

Supplier: Apollo Scientific

PC9637-5G
APOSPC9637-5GEA 284 EUR
APOSPC9637-5G
(ɑ,ɑ,ɑ,2,3,5,6-Heptafluoro-p-tolyl)hydrazine
(alpha,alpha,alpha,2,3,5,6-Heptafluoro-p-tolyl)hydrazine
Formula: C₇H₃F₇N₂
Melting Pt: 82…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00012368
CAS Number: 1868-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR