6-Hydroxy-4-(trifluoromethyl)nicotinic acid 95+%

Supplier: Apollo Scientific
Warning

PC9700-1G
APOSPC9700-1GEA 655 EUR
APOSPC9700-1G
6-Hydroxy-4-(trifluoromethyl)nicotinic acid 95+%
6-Hydroxy-4-(trifluoromethyl)nicotinic acid
Formula: C₇H₄F₃NO₃
Melting Pt: 302…305 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD06245493
CAS Number: 849020-87-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR