4-Bromo-2-chloro-5-fluorobenzoic acid

Supplier: Apollo Scientific
Warning

PC9723-1G PC9723-25G PC9723-5G
APOSPC9723-1GEA 21.9 EUR
APOSPC9723-1G APOSPC9723-25G APOSPC9723-5G
4-Bromo-2-chloro-5-fluorobenzoic acid
4-Bromo-2-chloro-5-fluorobenzoic acid
Formula: C₇H₃BrClFO₂
Melting Pt: 150…155 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00236589
CAS Number: 177480-81-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR