2-(Trifluoromethoxy)phenol 98%

Supplier: Apollo Scientific
Warning

PC9734-100G PC9734-25G PC9734-5G
APOSPC9734-100GEA 133 EUR
APOSPC9734-100G APOSPC9734-25G APOSPC9734-5G
2-(Trifluoromethoxy)phenol 98%
2-(Trifluoromethoxy)phenol
Formula: C₇H₅F₃O₂
Boiling Pt: 69…71 °C/60mmHg
Density: 1,332 g/cm³ (25 °C)
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00236324
CAS Number: 32858-93-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR