4-Amino-3-fluorobenzoic acid 97+%

Supplier: Apollo Scientific
Warning

Synonyms: 4-Amino-3-fluorobenzoic Acid

PC9752-1G PC9752-5G
APOSPC9752-1GEA 21.9 EUR
APOSPC9752-1G APOSPC9752-5G
4-Amino-3-fluorobenzoic acid 97+%
4-Amino-3-fluorobenzoic acid
Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Melting Pt: 212…218 °C
Storage Temperature: Ambient
MDL Number: MFCD01660374
CAS Number: 455-87-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR